United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
EW-18105-00
Bleach Spray System Kit, (includes six, 11-oz. bleach cartridges, two, 5000ppm spayer heads, and two, 11-oz. water bottles)
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
MSDS
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-18105-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Cleaning Supplies
>
Cleaning Solutions
>
Lab and General Purpose Cleaners
>
Bleach Spray System
Share
|
Bleach Spray System Kit, (includes six, 11-oz. bleach cartridges, two, 5000ppm spayer heads, and two, 11-oz. water bottles)
EW-18105-00
$88.00 USD/PACKAGE
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Eliminates hazard of bleach spills on skin or surfaces
This two-bottle fixed-ratio trigger sprayer automatically mixes and dilutes 5.25% bleach concentrate with tap water to make a 5000 ppm bleach disinfectant solution for use on hard, non-porous surfaces. The sprayer keeps the bleach separate from the water until you spray, keeping the 10% solution fresh and effective. Refill the water cartridge with tap water and lock in the disposable bleach cartridges and system is ready for use. Item 18105-00 kit includes six, 11-oz. bleach cartridges, two, 5000 ppm sprayer heads, and two 11-oz. water bottles.
Specifications
Product Type
Equipment Cleaner
Format
Spray
Packaging
6-11 oz bleach cartridges, 2-5000 ppm spayer heads; and 2-1 oz water bottles
Brand
DEARDORFF FITZSIMMONS CORP
Manufacturer number
77062
Model
77062
Accessories
Qty
/
Price
Availability
Activate
High-Level Chlorine Test Strips 50/PK
EW-18105-01
at
$36.00 USD/PACKAGE
In stock.
Add to Cart
Technical Resources
MSDS
MSDS 2 (135 B)
Related technical articles (Roll over for summary)
ChemicalsSpecial Ordering Information
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title