log in or register
Blue Dye Concentrate, Gallon
EW-00298-08
$108.00 USD/GALLON
  • Determine water flow in closed circuits or open channels
  • Biodegradable and nontoxic
  • Blue dye brightens ponds
Dyes diffuse rapidly in water, producing a vivid, easy-to-see color. Excellent for identifying diffusion and dispersion patterns, rates of flow, discharge locations, seepage areas, sea marking, and more. Use blue dye to brighten murky ponds and other landscaped waters.
Minimum detectable concentration of all dyes in clear water is 1 ppm. Available in tablets, liquid, or powder. Quick dissolving tablets are easy to handling with less mess than other forms. They are perfect for applications such as storm lines, cooling and plumbing systems, and sewer lines. Choose liquid for applications such as metered input, flow/dilution/disperation rate assessment, septic inspections, or fluoremetric dye tests. Powder is well-suited for large applications where liquid is undesirable such a cooling towers, lakes, rivers, and ponds, or power plant piping.
Product Type Dye Tracers
Gallons of water for 1 ppm 100,000
Applications Metered systems
Manufacturer number 106502G
Technical Resources
  • MSDS 1 (135 B)
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title