United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-86105-14
Body fluids clean-up kit
Click to enlarge
More Product Details
Specifications
Technical Resources
MSDS
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-86105-14
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
First Aid
>
Kits
>
Cole-Parmer Body-Fluid Clean-Up Kits
Share
|
Body fluids clean-up kit
EW-86105-14
$13.50 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Contains items mandated by OSHA and CDC.
Kit comes in plastic gasketed case which can be mounted on walls and easily removed for grab-and-go emergency use.
What's Included:
RED Z pouch, one pair of latex gloves, pick-up scoop with scraper, anti-microbial wipe, germicidal disinfectant surface wipe, wiper towel, biohazard waste bag, odor mask, easy-to-follow instructions.
Hot Tips:
To comply with OSHA 1910.151 for industry, and 1926.50 for construction, you must stock high-quality first aid supplies such as these Cole-Parmer kits in your workplace.
Technical Resources
MSDS 1 (135 B)
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title