Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Borosil Erlenmeyer Flask, glass, 1000 mL, 6/pk

Borosil  |   Mfr# FG4980-1000  |   Item# EW-61000-95

$87.50 USD/PACKAGE

Are you a reseller or buying in bulk? Call us today.


Usually ships in 6 days.
Qty
    Manufactured of borosilicate glass, flasks are provide thermal shock resistance, physical strength, and are resistant to chemicals.
    Capacity (mL) 1000
    Graduation range (low) 250
    Graduation range (high) 1000
    Material Glass
    Manufacturer number FG4980-1000
    Brand Borosil
    Qty/pk 6





    Technical Resources
     
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews