Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Botulinum Toxin Detection Kit, handheld assay, 10/bx
Item# EW-36108-14
$426.00 USD/BOX
Are you a reseller or buying in bulk? Call us today.
Usually ships in 9 days.
Qty
  • Quickly determine biological threat credibility
  • Results in 3- 30 minutes
  • Easy to read – two lines for positive; one line for negative
  • High Sensitivity and specificity
Accurately detect small, yet high threat amounts of biowarfare agents-without extensive sample preparation or additional expensive equipment. Test powder or liquid samples; kits are simple to use, results are quick and easy to read. Biohazard disposal bags and chain of custody labels are included for proper control and disposal or transport of the sample and test. Order simulation devices below for training purposes.
What’s included: 5" x 7" pouch includes all material to prepare and test a sample as well as package it for disposal or transport after testing.
Product Type Bacterial Test Kit
Tests for Botulinum Toxin
Qty/pk 10
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Do you have questions about this product?
Get answers from real customers and in-house experts with AnswerBox.
3 Questions | 3 Answers
Displaying questions 1-3
  • Q:
    What is the detection mechanism of this kit? Are there toxin detection kits for Listeria monocytogenes, Yersinia enterocolitica, Proteus mirabils and Staphylococcus aureus
    Asked on 12/11/2014 by Santosh IBSD
    1 answer
    • STAFF EXPERT
      A:
      Everything you need to perform the test is included in each kit. The test takes from 3-15 minutes to work and is a qualitative result. The test will only tell you that there is the Botulinum Toxin present, not how much is present. We do not offer kits for the other toxins listed.
      Answered on 12/11/2014 by Dan from Technical Support
  • Q:
    What are the values of sensitivity?
    Against that toxins is specificic?
    Asked on 2/20/2013 by Jose María from Valladolid, Spain
    1 answer
    • STAFF EXPERT
      A:
      The limit of detection for the two Botulinum toxins (A & B) are 33 ng/mL & 500 ng/mL. This test is not sensitive to the other toxin types (C-G).
      Answered on 2/22/2013 by Christina from Technical Support
  • Q:
    How many tests are there in each box of BADD.
    Asked on 12/28/2012 by Neeraja
    1 answer
    • STAFF EXPERT
      A:
      There are 10 tests per box.
      Answered on 12/28/2012 by Jen from Technical Support
Displaying questions 1-3
Do you have a question about this product? 
Product Reviews