BRAND Seripettor Economy Dispenser, 0.2 to 2 mL

Representative image only.
Mfr # 4720120 – Item # EW-07878-28
An economical choice for dispensing of many liquids in biological labs
  • Spring-loaded piston for one handed dispensing
  • Replaceable piston/cylinder cartridge to renew dispenser
  • Suitable for sodium azide solutions
  • 1 year warranty

PRODUCT OPTIONS

$174.00USD / Each

$89.50 - $162.00USD / Each

Time to recalibrate? Click on the calibration service to view details, then order.

SPECS & DESCRIPTION

Specifications & Description

  • Min Volume (mL)0.2
  • Max Volume (mL)2
  • Division (mL)0.04
  • Accuracy±1.2%
  • Reproducibility±0.2%
  • AutoclavableLower body only
  • Wetted PartsPP, PE, and EPDM
  • DescriptionSeripettor Economy Dispenser, 0.2 to 2 mL
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier

Key Features

  • Spring-loaded piston for one handed dispensing
  • Replaceable piston/cylinder cartridge to renew dispenser
  • Suitable for sodium azide solutions
  • 1 year warranty

More About this Item

The seripettor® is an economical dispenser suitable for many needs of the biology laboratory. An excellent choice for bases, buffers, and other aqueous solutions as well as dilute, non-oxidizing acids, and polar solvents. Wetted materials include PP, HDPE, and EPDM rubber. Not recommended for concentrated acids, non-polar solvents, or scintillation fluid. Accessories are available for dispensing of culture media or agar.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat