United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
3 >
Representative photo only
BrandTech Seripettor Economy Dispenser 1 0 to 10 mL
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07878-30
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17104-00
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
$66.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Dispensers
>
Bottletop Dispensers
>
BrandTech Seripettor Economy Dispensers
Share
|
BrandTech Seripettor Economy Dispenser, 1.0 to 10 mL
EW-07878-30
$138.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for sterile dispensing of culture media, buffers, and other biological solutions
These seripettor
®
dispensers offer rugged versatility and a low lifetime cost. You can easily remove and replace the dispensing cartridge without toolsreplacement cartridge is included. Use these dispensers with common laboratory reagents, including bases, biological solutions, dilute acids, polar solvents, and aqueous solutions. Not recommended for concentrated or oxidizing acids, or for nonpolar solvents. For sterile applications, order a MediaMate
kit separately. The kit allows you to dispense culture media, buffers, and other biological solutions. Each kit includes an autoclavable filling tube, two autoclavable 0.2 µm membrane filters, a valve block cap, and one pack of sterile replacement dispensing cartridges. Sterile replacement cartridges
Specifications
Product Type
Dispenser (Bottletop)
Volume (mL)
10
Accuracy
±1.2%
Division (mL)
0.2
Range (mL)
1 - 10
Reproducibility
±0.2%
Wetted parts
PP, PE, and EPDM
Includes
Autoclavable filling tube, 2 autoclavable 0.2 µm membrane filters, a valve block cap, and 1 pack of sterile replacement dispensing cartridges
Autoclavable
Lower body is autoclavable; pump assembly and cartridge are not
Brand
BrandTech
Manufacturer number
4721140
Model
4721140
Accessories
Qty
/
Price
Availability
Replacement Cartridges for BrandTech Seripettor Economy Dispenser, 1.0 to 10 mL, Pack of three
EW-07878-34
at
$44.50 USD/PACKAGE of 3
Usually ships in 7 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
EW-17104-00
at $66.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title