United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
6 >
Representative photo only
Brinkmann ChemSaver Dispenser 1 5 mL
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07932-01
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17104-00
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
$66.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Dispensers
>
Bottletop Dispensers
>
Brinkmann ChemSaver Dispensers
Share
|
Brinkmann ChemSaver Dispenser 1-5 mL
EW-07932-01
$588.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Unique PTFE “wiping” piston seal eliminates reagent residue in the cylinder and prevents “freezing” due to piston swelling and reagent crystallization
Safety recirculating valve channels reagent back into bottle during priming
Precise graduation scale allowing rapid volume setting
Telescopic filling tube adjusts to fit most reagent bottles
Dispenser rotates 360 degrees for easy and safe dispensing
Fully autoclavable assembly—no need to loosen valve or disassemble
Discharge tube surrounded by support sleeve to restrict movement and improve safety
Volume adjustment sleeve also rotates 360 degree for maximum visibility at all times
Fine volume adjustments for working with viscous or high-density liquids
These dispensers feature a smooth pump-action piston that draws and delivers precise volumes time after time. During delivery, a "wiping" seal of PTFE completely displaces reagent in the cylinder without leaving residue behind. This eliminates "freezing" due to piston swelling and reagent crystallization. Recirculating safety valve channels reagent back into bottle during priming.
Accuracy and precision values given for maximum volume
Brinkmann and Brinkmann ChemSaver—Reg TM Brinkmann Instruments, Inc.
Specifications
Product Type
Dispenser
Volume (mL)
5
Accuracy
±0.5%
Division (mL)
0.1
Range (mL)
1 - 5
Reproducibility
≤ 0.1%
Wetted parts
Borosilicate glass, ETFE, platinum-iridium, PP, and PTFE
Includes
Telescopic filling tube, wrench, & 3 adaptors (28, 38, & 45 mm bottles)
Autoclavable
Yes
Brand
Brinkmann
Manufacturer number
22 22 100-1
Model
22 22 100-1
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pipettors, with 10 measurements at 3 volumes
EW-17104-00
at $66.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title