log in or register
CALgrafix Process Monitoring and Configuration Software (other license can be added)
EW-89820-81
$199.00 USD/EACH
Usually ships in 11 days.
Qty
  • Process monitoring and configuration software
  • No design work required
CALgrafix is a cost-effective process monitoring and controller configuration software, which can be used with Cal Control or other brand controllers. Ideal in applications where there is a requirement to monitor and record temperature or other process values. The software is designed to control all functions of multiple controllers directly from your computer. System installations is fast and easy—simply place controllers wherever you need them and connect them to a central computer with the Windows® based software installed. CALgrafix software features datalogging, charting, alarm indication, instrument setting cloning, controller configuration/programming, and a virtual instrument display. Vast networking possibilities are available through the OPC client/server architecture. Up to 128 controllers can be controlled per each RS-485 network, and multiple networks can be monitored by one PC running CALgraphix software.
Product Type Temp control software
Brand Cal Control
Model 1003GB000
Manufacturer number 1003GB000
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title