Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register

CHEMets K-8012 Colorimetric Phenol Test Kit

CHEMets  |   Mfr# K-8012  |   Item# EW-00290-30

$77.25 USD/EACH
$69.52 USD/EACH

Are you a reseller or buying in bulk? Call us today.

Usually ships in 4 days.

  • CHEMets test kits are portable and self-contained
  • Vacuum sealed reagents and handy comparator chart for quick, visual results
CHEMETS colorimetric test kits let you visually compare the color of your sample to standards using a comparator. Just break tip of ampoule in cup (included) filled with sample. Vacuum pulls sample into ampoule automatically; gently swirl to mix. Handy color comparator (included) provides accurate visual analysis. Vacuum packing keeps reagents active indefinitely (see footnotes).
What's included: a polypropylene case with instructions, all necessary apparatus, and reagent vials for 30 tests (except detergent test kit, which has 20 tests). Refill kits contain 30 vials each (detergent refill kit has 20 vials).
Product Type Water Quality Test Kits
Range 0 to 1, 0 to 12 ppm
Test Phenol
Model K-8012
Qty/ea 1
Manufacturer number K-8012
Brand CHEMets


at $35.25 USD/BOX of 30

In stock.

Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers

    Do you have questions about this product?

    Get answers from real customers and in-house experts with AnswerBox.

    2 Questions | 2 Answers
    Displaying questions 1-2
    • Q:

      What is the minimum sample amount needed for the test. Where can I find the manual for detail procedure. Can we use it on very turbid samples with cell debries.
      Asked on 6/22/2016 by Sonali

      1 answer



        You would need 25 mL to fill up the tube to the correct mark. The procedures come in the kit

        Answered on 8/2/2016 by Anthony from Technical Support
    • Q:

      Would this product register the presence of tert-Butylphenol if the measurement range was accurate?
      Asked on 11/30/2011 by Jay from St Louis, MO

      1 answer



        We will need to know if the tert-butylphenol is o-tert-butyl, m-tert-butyl, or p-tert-butyl (or 2-, 3- or 4-tert-butylphenol). If it is p- or 4-, this kit will not be able to measure it. If it's o- or m- (2- or 3-), the kit will probably measure it but it is not confirmed how sensitive the kit will be, as this kit has not been tested experimentally with tert-butylphenols.

        Answered on 11/30/2011 by Rita from Customer Service
    Displaying questions 1-2

    Do you have a question about this product? 

    Product Reviews
    Customers Who Bought This Item Also Bought