United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
4 >
Representative photo only
CINC V2 Explosion Proof Centrifugal Separator Extractor 220 VAC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-17315-02
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Fluid & Liquid Handling
>
Centrifugal Separation
>
CINC V2 Centrifugal Multipurpose Separator/Extractors
>
CINC V2 Centrifugal Multipurpose Separator/Extractors
Share
|
CINC V2 Explosion-Proof Centrifugal Separator/Extractor, 220 VAC
EW-17315-02
$7,630.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Mix, react, and/or separate liquid-liquid mixtures in a single device
Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
Heavy-duty SS componentry can be quickly cleaned and inspected (suitable for CIP and autoclavable)
Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Users find this design to be superior to most disk-stack style centrifuges, gravity-based decanters, coalescer and DAF type separators, mixer/settler systems, and extraction columns. It is suitable for separation of liquids with specific gravity differences of 0.05 or better and can process immiscible droplet sizes that are up to 10 ìm. Users may interchange weirs to accommodate for liquids of varying specific gravity and viscosity. A second inlet port allows the introduction (down to 20 mL/min) of mixing additives to the liquid for extraction, reaction and washing.
What's Included:
centrifugal chamber, 1⁄8-hp electric motor with variable speed controller, high- and low-mix bottom plates plus ten weirs (ranging 0.825" to 1.050" dia), and a CD-ROM including weir selection software, dynamic-image instruction package, and maintenance guide. Explosion-proof motors add a 50-ft power cable and a hazardous environment multipin connector.
Specifications
Product Type
Centrifugal Separator
Wetted parts (rotor head and sleeve O-rings)
PTFE-encapsulated Viton®
Motor type
Explosion-proof
Refrigerated
No
Wetted parts (Housing)
316L stainless steel
Power (VAC)
220
Fitting
3/8" NPT(M)
Wetted parts (o-ring)
PTFE
Brand
CINC
Dimensions
9"W x 25"H x 9"D (22.8 cm x 63.5 cm x 22.8 cm)
Accessories
Qty
/
Price
Availability
Spare Parts Kit, contains common wear parts
EW-17315-08
at
$540.00 USD/SET
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Gulf Oil Spill Cleanup Puts Spotlight on Centrifugal Multipurpose Separators/Extractors
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title