Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

CINC V2 Standard Centrifugal Separator/Extractor, 220 VAC

EW-17315-00
$7,125.00 USD/EACH

  • Mix, react, and/or separate liquid-liquid mixtures in a single device
  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Heavy-duty SS componentry can be quickly cleaned and inspected (suitable for CIP and autoclavable)
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations
Users find this design to be superior to most disk-stack style centrifuges, gravity-based decanters, coalescer and DAF type separators, mixer/settler systems, and extraction columns. It is suitable for separation of liquids with specific gravity differences of 0.05 or better and can process immiscible droplet sizes that are up to 10 ìm. Users may interchange weirs to accommodate for liquids of varying specific gravity and viscosity. A second inlet port allows the introduction (down to 20 mL/min) of mixing additives to the liquid for extraction, reaction and washing.
What's Included:
centrifugal chamber, 1⁄8-hp electric motor with variable speed controller, high- and low-mix bottom plates plus ten weirs (ranging 0.825" to 1.050" dia), and a CD-ROM including weir selection software, dynamic-image instruction package, and maintenance guide. Explosion-proof motors add a 50-ft power cable and a hazardous environment multipin connector.
Product Type Centrifugal Separator
Wetted parts (rotor head and sleeve O-rings) PTFE-encapsulated Viton®
Motor type Standard
Refrigerated No
Wetted parts (Housing) 316L stainless steel
Power (VAC) 220
Fitting 3/8" NPT(M)
Wetted parts (o-ring) PTFE
Brand CINC
Dimensions 9"W x 25"H x 9"D (22.8 cm x 63.5 cm x 22.8 cm)

Accessories
Qty/Price
Availability
Free Cinc Cdrom
EW-00100-17

at Free

In stock.

at $540.00 USD/SET

In stock.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title