Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

CONTEC Prewetted Cleanroom Wipes, 30% DI water/70% IPA

EW-33675-02
$278.00 USD/CASE

    Prewetted wipes are saturated in a blend of DI water and isopropyl alcohol (IPA). Wipes are easily dispensed through a center-pull canister, keeping your workplace safe. Case includes twelve rolls of 100 wipes.


    Note: Not available for sale to the state of California


    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Wipes
    Description 30% DI water/70% IPA
    Quantity 12 rolls of 100 wipes
    Compliance Class 1000
    Brand Contec
    Manufacturer number SAT-C3-7030
    Model SAT-C3-7030





    Technical Resources
     
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title