United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
EW-07595-47
Cable assembly for remote control; 25-ft (7.9 m); model 07523-95 with DB25 connector is shown.
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07595-47
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Masterflex Peristaltic Pumps
>
Masterflex Tubing and Pump Accessories
>
Remote Control Cables, Connectors, and Accessories
>
Remote Control Cables
Share
|
Cable Assembly for Remote Control; DB9(M) connector and 25-ft (7.9 m) cable with stripped-wire ends.
EW-07595-47
$150.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Cables include noted terminal connector and cable with stripped wire ends for connection to PLC or other control interface
Use cable with DB9 male connector for remote control of Masterflex L/S 07557-00 and -10, the L/S 07528-series and the Masterflex I/P 07591-20 drives
Specifications
Brand
Masterflex
Manufacturer number
07595-47
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Reducing Pulsation
Peristaltic Tubing Pumps in Dispensing Applications
Show All...
Masterflex Tubing: Questions and Answers
Masterflex Tubing Pump Systems FAQs
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Peristaltic Pumps Not Just for Labs Anymore
Tubing Selection Guide For Viscous Fluids
Pumping Abrasives
Pumping Viscous Fluids
Peristaltic Pumps Offer Custom Fluid Solutions
Peristaltic Pumps Offer Economical Multichannel Pumping
Reducing Pulsation in Peristaltic Pumping Systems
Pumping Abrasives
Masterflex Systems: Questions and Answers
High-Performance Masterflex Pump Head
Six Ways to Simplify Fluid Transfer
Masterflex Pump FAQs
Masterflex Tubing Formulations
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title