Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Cannon standard constant temperature bath, 120 VAC

EW-98935-00
$5,559.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty

  • Take up to seven viscosity measurements at one time
  • Stainless steel cooling coil lets you operae below or slightly above ambient temperature
  • Temperature selection switch lets you quickly select from 10 popular temperature settings
  • Motor-driven stirrer ensures uniform temperature throughout bath
  • Ideal for kinematic viscosity measurements per ASTM D 445
Control the viscosity of your samples with these Cannon® constant-temperature baths. These unique baths hold up to seven glass capillary viscometers and two thermometers (holders are required—order separately). See-through glass chamber lets you continuously view samples.
All units feature a control circuit that provides proportional temperature control and an accuracy of ±0.01°C from 20 to 100°C (±0.03°C above 100°C for the high-temperature deluxe models). Select any temperature within the range or select from ten preset temperatures. Fine tuning dial lets you adjust this value to within 0.01°C of your target temperature. Multiple safety features include overtemperature, low-level, and sensor disconnection cutoffs.
Chamber is made of clear Pyrex® and includes a reflective stainless steel baffle in the center; the deluxe model also includes two fluorescent lamps on both sides. Housing is constructed of corrosion-resistant epoxy-coated heavy aluminum with insulated stainless steel cover; includes stainless steel cooling coil.
Fill bath with deionized water or oxidation-inhibiting bath oil for high-temperature applications. Order bath oil and other accessories separately, and thermometers from our Temperature section.
Product Type Viscosity Bath
Bath type Viscosity Water Bath
Capacity (Liters) 10
Min temperature (º C) 20
Max temperature (º C) 100
Temp control Proportional
Temp sensor Stainless steel-encased thermistor
High-temperature cutoff Adjustable within range
Temp setting Analog
Low-level cutoff Yes
Wetted materials Stainless steel and glass
Accuracy ±0.01ºC to 100ºC
Power (VAC) 120

Accessories
Qty/Price
Availability

at $48.00 USD/EACH

In stock.

at $28.00 USD/EACH

In stock.

at $48.00 USD/EACH

Call 1-888-358-4705 for availability.

at $29.00 USD/EACH

Call 1-888-358-4705 for availability.

at $182.00 USD/EACH

Call 1-888-358-4705 for availability.

at $182.00 USD/EACH

Call 1-888-358-4705 for availability.

at $31.00 USD/EACH

In stock.

at $187.00 USD/EACH

Call 1-888-358-4705 for availability.

at $164.00 USD/EACH

Call 1-888-358-4705 for availability.

at $958.00 USD/EACH

Call 1-888-358-4705 for availability.
Bath oil, 6 gallons
EW-98935-52

at $266.00 USD/EACH

In stock.

at $1,540.00 USD/EACH

Call 1-888-358-4705 for availability.

at $219.00 USD/EACH

In stock.

at $219.00 USD/EACH

Call 1-888-358-4705 for availability.

at $219.00 USD/EACH

Call 1-888-358-4705 for availability.
Viscosity standard S3
EW-98944-66

at $163.00 USD/EACH

In stock.
Viscosity standard S6
EW-98944-68

at $163.00 USD/EACH

In stock.

at $290.00 USD/EACH

In stock.

at $163.00 USD/EACH

Usually ships in 10 days or sooner.

at $163.00 USD/EACH

In stock.

at $163.00 USD/EACH

In stock.

at $163.00 USD/EACH

In stock.

at $163.00 USD/EACH

In stock.

at $163.00 USD/EACH

In stock.

at $163.00 USD/EACH

In stock.

at $163.00 USD/EACH

In stock.

at $163.00 USD/EACH

Call 1-888-358-4705 for availability.

at $163.00 USD/EACH

Call 1-888-358-4705 for availability.

at $163.00 USD/EACH

In stock.

at $163.00 USD/EACH

Call 1-888-358-4705 for availability.

at $163.00 USD/EACH

Call 1-888-358-4705 for availability.




Technical Resources
  • Instruction Manual.pdf
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.

    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title