United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
7 >
EW-83090-50
Capillary Cartridge Replacement for picoSpin-45 Benchtop NMR Spectrometer
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-83090-50
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Spectroscopy and Spectrophotometers
>
Nuclear Magnetic Resonance (NMR) Spectroscopy
>
picoSpin-45 Benchtop NMR Spectrometer Accessories
Share
|
Capillary Cartridge Replacement for picoSpin-45 Benchtop NMR Spectrometer
EW-83090-50
$672.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Check the fittings on your unit to decide if 1/32" or 1/16" accessories are right for you.
The capillary cartridge probe is a sub panel assembly containing the NMR RF coil and shim coils. Replacement of the capillary cartridge is necessary should the 300 µm microbore sample delivery tubing become clogged. To minimize downtime it is also advisable to keep a spare cartridge in stock. The picoSpin-45 Capillary Cartridge comes complete with the tools needed to remove and install the capillary cartridge sub panel. First generation capillary cartridges come standard with 1/32-inch fittings. Second generation cartridges come with 1/16-inch fittings.
In order to know if you have 1/16-inch or 1/32-inch fittings, look at the inlet fitting of your picoSpin-45. If you see two stainless hex nuts, you have 1/32-inch fittings. If you see one stainless hex nut, you have 1/16-inch fittings.
In general, capillary cartridges shipped before January 2013 have 1/32-inch fittings. Capillary cartridges shipped after January 2013 generally will have 1/16-inch fittings. Please contact our applications specialists if you need further assistance.
Specifications
Product Type
Capillary Cartridge
Brand
picoSpin
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Seeing the Light: An Overview of Visible and UV-VIS Spectroscopy
Spectrophotometer Selection Guide
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title