Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Captair Pyramid Glove Bag

EW-33666-50
$380.00 USD/EACH

Usually ships in 6 days or sooner.
Qty

  • Easily set up a controlled environment wherever you need it
  • Protects you from hazardous materials
  • Saves lab space
Set up this light-weight pyramid glove bag to provide a controlled environment for user protection and still-air materials handling. Easily transport the lightweight, self-supporting pyramid to any location and assemble in minutes.
Unit features an 18-1/2” (47 cm) side opening for introduction of materials, a zip lock to ensure air-tight sealing, and a port to exhaust/purge gas. Pyramid is constructed from transparent 250m PVC with frame and 0.2 mm thick butyl gloves. Glove bag is collapsible for easy storage, and can be cleaned with a mild cleanser for continued use. Glove bag cannot be autoclaved.
What's Included:
1/8" ID coupler body valved fitting
Chamber size 30 in W x 28.5 in H x 22 in D
Product Type Glove Bags
Side door dimensions 18.5 in (47 cm)
Dimensions 76.2 x 72.4 x 55.9 cm
Brand Captair
Manufacturer number 2200ANM
Model 2200ANM





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title