United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-01506-35
Cartridge,research +
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-01506-35
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Water Purification
>
Water Purification Cartridges
>
Cole-Parmer Ion-X-Changer Filter Cartridges
>
Cole-Parmer Ion-X-Changer Filter Cartridges - Research cartridge
Share
|
Cartridge,research +
EW-01506-35
$110.00 USD/EACH
Quantity Discounts
Order
Discount
Price
[X]
6
10%
$99.00/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Purify water up to 15 MΩ-cm
Design your system with any combination of four different cartridges
Our four grades of disposable Ion-X-Changer filter cartridges withstand water pressures as high as 100 psi for safe hookup to any laboratory tap water system. Cartridge resins turn amber to indicate when cartridges need to be changed. All cartridges must be used with mounting brackets (order separately).
Suggested System Combinations
A) Adsorber/Universal System combination produces water free of all ionized constituents except free CO
2
and silica.
B) Two Universal and Two Research Cartridges operating in tandem increase deionization capacity by 15%. Recommended for use with water that has a mineral salt concentration of 10 grains per gallon or more. When cartridges are mounted in pairs, use until the left cartridge is spent. Then, shift the right cartridge to the left side and replace the right side with a new cartridge. High water quality is retained.Research cartridge produces water ionically equivalent to triple-distilled water, removing all ionized minerals down to a level of 4 ppb or less as well as CO2 and silica. When used in tandem, these cartridges produce water with a resistivity of 15 M?-cm or greater. Capacity is 1000 grains expressed as CaCO
3
.
Specifications
Product Type
Water Purification Cartridge
Applications
Produces water ionically equivalent to triple-distilled water
Capacity (grains)
1000 as CaCO
3
Dimensions (" L)
18.75
Dimensions (" Dia)
3.25
Temperature (Max)
75° F (23.9° C)
Flow rate (Max)
7.2 GPH (27.3 L/hr)
Inlet pressure (Max)
100 psi (6.9 bar)
Inlet pressure (min)
5 psi (0.3 bar)
Manufacturer number
D502122
Model
D502122
Accessories
Qty
/
Price
Availability
Mounting bracket kit for Ion-X-Changer Filter Cartridges
EW-01506-60
at
$190.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Instruction Manual (564 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Water Purification
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Basics of Cartridge Filtration
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title