Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Casella USA dBadge Micro Noise Dosimeter Kit, 5pk

EW-86536-03
$6,900.00 USD/EACH

Usually ships in 8 days.
Qty

  • More convenient than the traditional noise dosimeter with remote cable microphone
  • Lightweight less than 2.5 oz
  • Small size—only 2.8" long, 1.7" wide and 2" tall
  • Quick charge in less than 1.5 hours from completely empty
  • Stores all results for later review
  • Calculates all the popular noise dose indices
  • Time history profiling of 4 results at fixed 1 min intervals
  • Wide dynamic range from 65 to a full 140 dB
  • Measures simultaneously with 3 dB and 5 dB exchange rates
  • Lockable in operation mode
  • Direct automatic calibration with standard acoustic calibrator (1⁄2" cavity)
  • Intrinsically safe model allows use in hazardous area such as petrochemical plants. This unit has worldwide ATEX approval
The CEL350 has all the advantages of earlier badge designs, such as small size and light weight, and yet it still provides all of the benefits that users have come to expect from a personal noise dosimeter. The CEL350 is fully featured and able to satisfy all current noise measurement protocols during a single run—an ideal instrument for both experienced and new users of dosimeters. Unit stores all results for later investigation and reporting. Easy, push-button setup.
What's Included:
110/2 calibrator, clip mounting set, windscreen, 3 way charger unit (for 1 to 3 dosimeter kits), 3 way slave charger (for kits of 4 or more), Windows PC software package, infra red interface module, RS-232 to USB adapter, and carrying case.
Product Type Noise Dosimeters
Description Kit containing 5 x CEL-350 MND, calibrator, case, master charger,power supply, slave charger, software, CMC39 adapter and RS-232 to USB adapter
Criterion level 70 to 90 dB (in 1 dB steps)
Exchange rate 5 and 3 dB simultaneously
Threshold level 70 to 90 dB (in 1 dB increments)
Weighting A for rms values plus Z and C for peak measurements
Instrument wt < 2.5 oz (68 g)
Resolution (dB) 0.1
Ouput signal RS-232
Accuracy ANSI Si.25, IEC61252 1997 Personal sound exposure meters
Function Leq, Lavg, Lmx, Lmn, LCpk, LZpk, LE, TWA, LEP, d, Dose%, Pdose%
Display LCD
Dimensions Badge: 2.75 in W x 1.75 in H x 2 in D; Reader unit: 3.25 in W x 6.2 in H x 1.5 in D
CE Compliance Yes
Brand Casella
Manufacturer number CEL-350/K5
Model CEL-350/K5

Accessories
Qty/Price
Availability

at $1,495.00 USD/EACH

Call 1-888-358-4705 for availability.
Calibrations
Qty/Price
Availability

at $150.00 USD/EACH
Usually ships in 8 days.




Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.

See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title