United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
8 >
Representative photo only
Cast iron float and thermostatic steam trap in line horizontal configuration 3 4 NPT F 15 psig
Click to enlarge
More Product Details
Specifications
Accessories
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07600-46
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Process Equipment & Instrumentation
>
Steam Handling
>
Spirax Sarco Thermostatic Steam Traps
>
Cast iron float and thermostatic steam traps (in-line horizontal configuration)
Share
|
Cast iron float and thermostatic steam trap (in-line horizontal configuration); 3/4" NPT(F), 15 psig
EW-07600-46
$261.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Drain condensate as quickly as it forms while removing air and other noncondensable gases
Float and thermostatic steam traps contain a float valve mechanism which modulates to discharge condensate continuously at steam temperature, while non-condensable gases are released by a separate internal balanced pressure thermosatic air vent. Stainless steel internal parts can be serviced without disturbing the piping connections.
Typical applications:
Air heating coils
Heat exchangers
Steam main drip stations
Small process equipment, particularly when controlled by a modulating temperature control valve
Replacement of less efficient inverted bucket traps on heating equipment
Specifications
Product Type
Steam handling fittings
Max operating pressure
15 psig
Material
Cast iron
Max temperature
45°F (25°C) of superheat
NPT(F)
0.75
Brand
Spirax Sarco
Manufacturer number
FTI-15 3/4" 66669
Model
FTI-15 3/4" 66669
Accessories
Qty
/
Price
Availability
General-Purpose Anti-Seize Thread Compound, 8-oz jar
EW-08809-22
at
$16.00 USD/EACH
In stock.
Add to Cart
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title