United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
3 >
EW-29300-00
Cell culture flask, 500 ml
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-29300-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Fermentation
>
Fermentation Components & Accessories
>
Mini Suspension Culture Systems
>
Cell Culture Spin Flasks
Share
|
Cell culture flask, 500 ml
EW-29300-00
$355.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Perfect for small scale operations, or quick tests of your fermentation process, these spin flasks are designed for microcarrier and suspension cultures, such as insect cells, hybridomas, and adapted cell lines.
Use the 45-mm sidearms as air vents, inoculation ports, or media inlet/outlet ports, or use sidearms to insert probes for pH, dissolved oxygen, or CO
2
measurement and control. Each spin flask features a bottom dimple to prevent cell crushing and improve circulation. Flasks are made of borosilicate glass, and include a PTFE and glass impeller assembly. Sidearms have PTFE-faced silicone liners. Top (center) cap has two liners: one silicone and one PTFE (to be used together). Entire spin flask unit is autoclavable. Additional sizes are available; please call our Applications Specialists for details.
Specifications
Material
Glass
Height (cm)
17.00
Accessories
Qty
/
Price
Availability
L/S Economy Pump System with Easy-Load II Pump Head, 115 VAC
EW-77910-20
at
$965.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Cole-Parmer Introduces Enhancements to Life Science Offering
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title