United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 4
of
6 >
Representative photo only
Centrifugal Magnetic Drive Pump with Enclosed Motor High Head Polypropylene 4 6 GPM 230 VAC
Click to enlarge
More Product Details
Specifications
Technical Resources
Flow Charts
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-72012-15
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Centrifugal Pumps
>
Centrifugal Pumps that are Magnetically Coupled (have a Magnetic Seal)
>
Centrifugal Magnetic Drive Pumps with Enclosed Motor, High-Head Polypropylene
Share
|
Centrifugal Magnetic Drive Pump with Enclosed Motor, High-Head Polypropylene, 4.6 GPM, 230 VAC
EW-72012-15
$362.00 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
Twin bearing system handles thrust loads for extended service life
Quiet, energy efficient, and compact
Internal cooling circuit reduces bearing temperature
Enclosed motor for use in harsh environments
All wetted parts are nonmetallic for excellent chemical resistance
High-head polypropylene pumps have a closed impeller design to significantly increase the output pressure. Polypropylene construction provides excellent chemical resistance in applications, such as filtration, where a high-head is needed.
What's Included:
A 6-ft (1.8 m) cord with stripped ends.
Specifications
Product Type
Magnetic Drive Centrifugal Pumps
Max flow rate (GPM)
4.6
Max flow rate (LPM)
17.4
Max head (ft)
36.1
Max head (m)
11.0
Max system pressure (psi)
24.2
Max system pressure (bar)
1.7
Motor (rpm)
3400
Motor (VAC)
230
Motor (Amps)
0.50
Motor (hp)
1/16
Wetted materials
Glass fiber-reinforced PP casings & impeller/magnet capsule
Alumina ceramic 99.5% spindle & thrust ring
Viton® O-ring
PTFE bearing
Max fluid temp (º F)
176
Max fluid temp (º C)
80
Duty cycle
Continuous
Connections (inlet)
5/8" hose barb
Connections (outlet)
5/8" hose barb
CE Compliance
Yes
Dimensions
9-5/8"L x 4-3/4"W x 5-1/8"H
(24.4 x 12.1 x 13 cm)
Technical Resources
CP_Flow_Curve_72012-15.pdf
Instruction Manual (42 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Centrifugal Pump Selection Guide
Gravity Feed Installation of Your Centrifugal Pump
How to Read a Pump Curve
Show All...
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title