United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
EW-06274-06
CENTRIFUGE TUBE ICE RACK COVER
Click to enlarge
More Product Details
Specifications
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-06274-06
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Laboratory Supplies & Consumables
>
Ice Buckets and Ice Pans
>
Centrifuge Tube Ice Tray, polypropylene
Share
|
Centrifuge Tube Ice Rack Cover
EW-06274-06
$18.00 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
For temperature stabilized benchtop ice storage
This heavy duty, all polypropylene rack provides efficient and extended cooling of twenty-eight 15 ml or 50 ml centrifuge tubes in a chilled bath. The deep bottom tray holds approximately 2 liters of ice flakes and water so that tubes can rest on the bottom of the tray for maximum stability and exposure to the bath. A flat perimeter rim provides space for labeling tape. The rack is steam autoclavable at 121°C (250°F) and stackable with identical racks when full or empty. To further protect tubes during transport or storage while also maintaining stackability, an optional polypropylene cover is available (order separately).
Specifications
Product Type
Centrifuge/Microcentrifuge Racks
Tube size
15 ml or 50 ml centrifuge tubes
Material
Polypropylene
Manufacturer number
F18905-0051
Model
F18905-0051
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title