United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
6 >
Representative photo only
Chemical Duty Transfer Pump with Kynar Head 18 GPM max flow
Click to enlarge
More Product Details
Specifications
Technical Resources
Flow Charts
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-07190-40
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Transfer Pump
>
Chemical-Duty Transfer Pumps
Share
|
Chemical-Duty Transfer Pump with Kynar Head, 18 GPM max flow
EW-07190-40
$1,029.00 USD/EACH
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Self-priming pumps with glass-filled polypropylene heads are capable of lifting water up to 10 feet
Pumps with Kynar
®
PVDF heads are ideal for ultra-high purity, chemical, and industrial applications
What's Included:
Conduit box for hardwiring.
Specifications
Product Type
Magnetic Drive Centrifugal Pumps
Max flow rate (GPM)
18
Max flow rate (LPM)
68.1
Max head (ft)
29
Max head (m)
8.8
Max system pressure (psi)
75
Max system pressure (bar)
5.2
Motor (phase)
1
Motor (rpm)
3450
Motor (VAC)
115/230
Motor (Hz)
50/60
Motor (Amps)
2.2/1.1
Motor (hp)
1/5
Motor (type)
TEFC
Wetted materials
Kynar®, 99.9% aluma oxide ceramic, PTFE encapsulated Viton®
Max fluid temp (º F)
200
Max fluid temp (º C)
93
Duty cycle
Continuous
Connections (inlet)
1" NPT(F)
Connections (outlet)
1/2" NPT(M)
Suction lift
10 ft H
2
O
Dimensions
11-3/8"L x 6-13/16"W x 6"H
(28.9 x 17.3 x 15.2 cm)
Brand
Kynar
Technical Resources
CP_Flow_Curve_07190-40.pdf
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title