Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Chemostat Tube for 15-Liter Fermentation/Bioreactor Systems
Item# EW-01919-46
$1,267.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Available for order
or call 1-888-358-4705 to order
Qty
    Chemostat tubes are used with a Masterflex® tubing pump (sold separately) to draw off harvested media or to add fresh media to maintain constant media level.
    Product Type Chemostat tube
    Qty/ea 1
    Brand Applikon
    Technical Resources
    Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456