United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 12
of
20 >
Representative photo only
Clear C FLEX Tubing 1 4 ID x 7 16 OD 25 ft pack
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-06422-11
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Tubing, and Hose
>
Tubing
>
Flexible Tubing
>
Clear C-FLEX Tubing
Share
|
Clear C-FLEX Tubing, 1/4"ID x 7/16"OD, 25 ft/pack
EW-06422-11
$63.00 USD/PACKAGE
Quantity Discounts
Order
Discount
Price
[X]
4
10%
$56.70/PACKAGE
10
15%
$53.55/PACKAGE
20
20%
$50.40/PACKAGE
In stock.
Qty
:
Add to Cart
Save to shopping list
Applications:
Pharmaceutical, biotech, and general-purpose lab uses requiring high performance. Economical alternative to platinum silicone.
Characteristics:
Exceptional tensile and tear strength, good biocompatibility, excellent chemical resistance, low gas permeability, and a smooth surface. Transparent.
Certification:
Product is manufactured in accordance with quality regulations CFR Title 21, Part 820 Quality System Requirements (GMP).
Temperature range:
-40 to 104°F (–40 to 40°C)
Sterilize:
Autoclave, ethylene oxide, or gamma irradiation.
Specifications
Product Type
Tubing
Working pressure (psi)
28
Maximum temperature (º F)
104
Minimum temperature (º F)
-40
Max temperature (º C)
40
Minimum temperature (º C)
-40
Tubing ID (in.)
1/4
Tubing ID (mm)
6.4
Tubing OD (in.)
7/16
Tubing OD (mm)
11.2
Wall thickness (in.)
3/32
Wall thickness (mm)
2.4
Max psi
28
Certifications
USDA, USP, GMP
Translucent
Yes
Sterilizable
Yes
Color
Translucent
Feet per pack
25
Tubing formulation
C-Flex
Brand
C-FLEX
Manufacturer number
190-940-001
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Cole-Parmers Bioconnect Program
Decrease bacterial growth in your tubin
The STAMP Tubing Selection Guide
Show All...
Tygon® Flexible Plastic Tubing to Tubing Solvent Sealing
Tubing Selection Guide
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title