Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
log in or register
Home  > 
Discontinued Items >
Sorry, this item is now discontinued.browse similar itemsor Contact us for more information.

Clenbuterol Hydrochloride (250 MG)

Item# EW-00022-OO

Call for Price

CAS Number 21898-19-1
Formula weight 313.66

Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews