Item Total = $0.00 
Items = 0 
View Cart
United States
1-888-358-4717 or 
Welcome,
log in or register
Sorry, this item is now discontinued. Please browse similar items.
or Contact us for more information.

Clenbuterol Hydrochloride (250 MG)

Item# WU-00022-OO

Call for Price

Discontinued
Qty
CAS Number 21898-19-1
Formula weight 313.66





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-888-358-4717
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews