United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 8
of
18 >
EW-01540-08
Representative Photo Only. Cole-Parmer
®
2-Way Direct Lift Solenoid Valve; normally closed valves 5/32 24VDC
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-01540-08
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Flow, Level & Valves
>
Valves
>
Solenoid Valves
>
Cole-Parmer 2- and 3-way Direct Lift Solenoid Valves
>
Cole-Parmer 2- and 3-way Direct Lift Solenoid Valves
Share
|
Cole-Parmer 2-Way Direct Lift Solenoid Valve; normally closed valves 5/32 24VDC
EW-01540-08
$180.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Ideal for use with high-purity or corrosive fluids
All wetted parts made of inert PTFE
Minimal dead volume reduces problems with trapped liquids
Rated for 26" Hg vacuum. Maximum fluid temperature is 158°F (70°C). All valves have 15" lead wires. Valves are compact and lightweightmount them almost anywhere. Valves with a 1/16" orifice have mounting holes at the bottom only and valves with a 3/32" and 5/32" orifice have mounting holes at the top and bottom.
Specifications
Product Type
Solenoid Electric Valves (actuated)
Watts
8
Max psi
18
Max temperature
158°F (70°C)
Orifice (in.)
5/32
Port size (NPT(F))
1/8
Response
30 msec
Max pressure (psi)
18
Power (VDC)
24
CE Compliance
Yes
Accessories
Qty
/
Price
Availability
CoolCube
Circuit
EW-01356-52
at
$44.00 USD/EACH
In stock.
Add to Cart
Mounting Clip
EW-01540-50
at
$3.50 USD/EACH
In stock.
Add to Cart
Mounting Flange for Cole-Parmer Solenoid Valves
EW-01540-55
at
$2.50 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Olfactometers and using your nose
Solenoid Valves and Viscous Fluids
Basics of Cartridge Filtration
Show All...
Valve Selection Guide
Electron zap turns flimsy plastic into sturdy shrink wrap
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title