United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
4 >
Representative photo only
Cole Parmer 304 SS Mechanically Coupled Pumps Self Priming 120 GPM or 20FT 3 4HP TEFC 1 phase
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-75505-71
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Centrifugal Pumps
>
Centrifugal Pumps that are Mechanically Coupled (have a Mechanical Seal)
>
Cole-Parmer 304 SS Mechanically Coupled Pumps, Self-Priming
Share
|
Cole-Parmer 304 SS Mechanically Coupled Pumps, Self-Priming, 120 GPM or 20FT, 3/4HP, TEFC, 1 phase
EW-75505-71
$665.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
304 stainless steel (SS) pump heads provide dependable performance and rugged continuous duty service with chemical and corrosion resistance
Ideal for food and pharmaceutical processing plants, as well as water and wastewater treatment centers
Close-coupled design saves space and simplifies installation and maintenance
Back pullout constructionassembly and overhaul of impeller and seal without distorting suction and discharge connections
Top centerline discharge and foot support under casing ensure self-venting and reduce misalignment from pipe loads
High-quality mechanical shaft seals and O-rings available for standard pumping requirementsalso available are optional materials for high-temperature and chemical-duty operation (contact an Application Specialist).
What's Included:
Conduit box for hard-wiring.
Specifications
Product Type
Mechanically Coupled Centrifugal Pumps
Max flow rate (GPM)
19
Max flow rate (LPM)
71.9
Max head (ft)
120
Max head (m)
36.6
Max system pressure (psi)
125
Max system pressure (bar)
8.6
Motor (phase)
1
Motor (rpm)
3450
Motor (VAC)
115/230
Motor (Hz)
60
Motor (Amps)
12.0/6.0
Motor (hp)
3/4
Motor (type)
TEFC
Wetted materials
304 SS pump head and impeller; SS shaft;
Type 21 mechanical seals (carbon vs. ceramic/Viton®);
and Viton® O-rings
Duty cycle
Continuous
Connections (inlet)
1-1/4" NPT(F)
Connections (outlet)
1" NPT(F)
CE Compliance
Yes
Dimensions
19-1/16" x 9-3/16" x 8-5/16"
(48.4 x 23.3 x 21.1 cm)
Brand
Cole-Parmer
Accessories
Qty
/
Price
Availability
Flange Kit for 1" and 1-1/4" Connections
EW-70725-70
at
$352.00 USD/EACH
Usually ships in 14 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Centrifugal Pump Selection Guide
Gravity Feed Installation of Your Centrifugal Pump
How to Read a Pump Curve
Show All...
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title