United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
EW-05802-14
CONTROLLER, PH/ORP 1/8 DIN
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-05802-14
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17106-20
NIST-Traceable Calibration; pH Meter
$115.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
pH Meters and Water Quality Products
>
Process pH and Water Quality Control
>
pH/ORP Control
>
1/8-DIN pH/ORP Control
>
Cole-Parmer pH/ORP/Temperature 1/8-DIN Controller and Transmitter
Share
|
Cole-Parmer 350 pH/ORP/Temperature 1/8-DIN Controller
EW-05802-14
$219.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Economical option with two SPDT relays or with isolated 4 to 20 mA output
Backlight LCD for clear measurement display
Automatic buffer recognition of US and NIST buffers
Universal power supply input from 100 to 240VAC
Easy-to-install 1/8-DIN compact plastic casing
Cole-Parmer pH/ORP controller and transmitter offer features found in more expensive models. Automatic buffer recognition, absolute and relative ORP measurements, and automatic temperature compensation ensure measurement accuracy.
Model pH/ORP 350 features two sets of programmable on/off relays for high/low control to activate an alarm, a pump, or another device.
Specifications
Product Type
Controller
Range (pH)
0.00 to 16.00
Range (Temperature)
10 to 120°C
Range (mV)
±1999
Accuracy (pH)
±2
Accuracy (Temperature)
±1% + 1 digit
Accuracy (mV)
±2
Resolution (pH)
0.01
Resolution (Temperature)
0.1
Resolution (mV)
1
Output (Current)
Isolated 4 to 20 mA
Dimensions
3 2/3" x 1 3/4" x 4 1/4"
Power
100 to 240 VAC
Housing
1/8-DIN
Brand
Cole-Parmer
Accessories
Qty
/
Price
Availability
Cole-Parmer self-cleaning pH electrode, Ryton housing, 1 kΩ RTD, 10-ft cable, BNC
EW-27003-24
at
$176.00 USD/EACH
Call 1-888-358-4705 for availability.
Add to Cart
Cole-Parmer self-cleaning ORP electrode, Ryton housing, platinum band, 10-ft cable, BNC
EW-27003-50
at
$187.00 USD/EACH
In stock.
Add to Cart
Cole-Parmer Tuff-Tip, Semi-Domed 1" In-Line or Submersible Graphite Tipped pH Electrode, 1 KOhm RTD(Spade Lugs), BNC
EW-27020-66
at
$248.00 USD/EACH
Usually ships in 12 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; pH Meter
EW-17106-20
at $115.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
pH/ORP Control Systems Selection Guide
Typical Problems in Industrial pH Measurement & Control
NEMA/IEC Enclosure Ratings
Show All...
Taking Control of Your pH!
Tech Challenge: Faulty pH Readings
Testing the pH of Wastewater Samples
Testing the pH of Soil Samples
Electron zap turns flimsy plastic into sturdy shrink wrap
Basics of Cartridge Filtration
Bitter Brew
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title