dummy
EW-04711-70
Call 1-888-358-4705 for availability.
Qty:
- Process batch samples as small as 250µL or as large as 19 L/hr using optional continuous flow cell
- Exclusive energy (joules) monitor provides the most accurate method to reproduce results
- On/off independent pulser helps prevent overheating
- User-friendly menu-driven prompts provide intuitive guidance through all functions
Our ultrasonic processors are capable of delivering 500 or 750 watts at 20 kHz. The 500-watt models are recommended for smaller volumes, and for probes of 1/2-inch or less. The 750-watt models are recommended for larger volumes, 3/4-inch probes, and the continuous flow cell and cup horn. Process batch samples as small as 250 µL or as large as 19 L/hr using the continuous flow cell 04710-91 (order separately). The sealed converter protects the unit from humidity, dust, dirt, and fumes. Units monitor both power (watts) and energy (joules), and feature 100% automatic tuning. The independent on/off pulser ensures safe treatment of temperature-sensitive samples while processing at high intensity and controls on/off cycles from 1 to 59 seconds. The microprocessor-based timer controls your processes from 1 second up to 10 hours. Timer monitors both the elapsed time and duration of processing.
The tactile keypad, menu-driven display, and simple function keys combine for a user-friendly system. All programmed control parameters are displayed on the easy-to-read screen so you can continuously monitor your process. Adjust cavitation energy intensity delivered to your samples with the variable amplitude control. The amount of power being delivered to the probe is displayed both digitally and as a bar graph. View selected output level on the screen.
What's Included:
converter, tool kit, foot switch connection, 13-mm dia (½") probe 04710-31 with replaceable tip, and a 6-ft cord with plug.
| Product Type |
Ultrasonic Processor |
| Description |
Cole-Parmer® 500-Watt Ultrasonic Homogenizer, 115 VAC |
| Min sample size (mL) |
0.25 |
| Max sample size (mL) |
19000 |
| Frequency (kHz) |
20 |
| Watts |
500 |
| Power (VAC) |
115 |
| Power (Hz) |
50/60 |
| CE Compliance |
Yes |
| Dimensions (" W) |
7.5 |
| Dimensions (" H) |
8.5 |
| Dimensions (" D) |
13.5 |
| Display |
LCD |
| Manufacturer number |
VC505-115V/60HZ |
| Brand |
Cole-Parmer |
at $665.00 USD/EACH
In stock.
at $750.00 USD/EACH
Call 1-888-358-4705 for availability.
at $144.00 USD/EACH
Usually ships in 12 days.
at $138.00 USD/EACH
Usually ships in 8 days or sooner.
at $100.00 USD/EACH
Call 1-888-358-4705 for availability.
at $520.00 USD/EACH
Usually ships in 11 days.
at $60.25 USD/EACH
In stock.
at $67.00 USD/EACH
In stock.
at $542.00 USD/EACH
Usually ships in 9 days.
at $542.00 USD/EACH
Usually ships in 26 days.
at $542.00 USD/EACH
Usually ships in 15 days.
at $72.00 USD/EACH
In stock.
at $72.00 USD/EACH
Usually ships in 15 days.
at $72.00 USD/EACH
Usually ships in 15 days.
at $542.00 USD/EACH
Usually ships in 17 days.
at $542.00 USD/EACH
Usually ships in 20 days.
at $665.00 USD/EACH
Usually ships in 7 days.
at $1,396.00 USD/EACH
Usually ships in 56 days.
at $4,030.00 USD/EACH
Usually ships in 21 days.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.