log in or register
Cole-Parmer 500-Watt Ultrasonic Homogenizers with Temperature Controller, 230 VAC
EW-04711-35
$4,480.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Process batch samples as small as 250 µL or as large as 19 L/hr using optional continuous flow cell
  • Integral temperature controller and memory prevents harmful overheating of the sample by terminating the ultrasonics when the sample reaches a predetermined temperature limit
  • Exclusive energy (joules) monitor provides the most accurate method to reproduce results
  • On/off independent pulser helps prevent overheating
  • User-friendly menu-driven prompts provide intuitive guidance through all functions
These ultrasonic processors have the same features as the models above plus an integral temperature controller and memory. The temperature controller prevents harmful overheating of the sample by terminating the ultrasonics when the sample reaches a predetermined temperature limit (select from 1 to 100ºC); order temperature probe separately below. The time-saving memory stores up to ten procedures to facilitate protocol duplication, automate repetitive tasks, and eliminate user-to-user method variability for consistent reproducibility.
What's Included:
converter, tool kit, foot switch connection, 1⁄2" (13-mm dia) probe 04710-31 with replaceable tip, and a 6-ft cord with plug.
Product Type Ultrasonic Processor
Description Cole-Parmer 500-Watt Ultrasonic Homogenizers with Temperature Controller, 230 VAC
Min sample size (mL) 0.25
Max sample size (mL) 19000
Frequency (kHz) 20
Watts 500
Power (VAC) 230
Power (Hz) 50/60
CE Compliance Yes
Dimensions (" W) 7.5
Dimensions (" H) 8.5
Dimensions (" D) 13.5
Display LCD
Manufacturer number VCX500-230V/50HZ
Brand Cole-Parmer
Accessories
Qty/Price
Availability
Cup horn; 2-1/2" dia
EW-04710-38
at $1,120.00 USD/EACH
Call 1-888-358-4705 for availability.
at $55.50 USD/EACH
Usually ships in 10 days.
at $665.00 USD/EACH
In stock.
at $315.00 USD/EACH
Call 1-888-358-4705 for availability.
Cooling cell, 300 mL
EW-04710-89
at $225.00 USD/EACH
Call 1-888-358-4705 for availability.
Continuous Flow Cell
EW-04710-91
at $750.00 USD/EACH
Call 1-888-358-4705 for availability.
at $144.00 USD/EACH
Usually ships in 12 days.
at $138.00 USD/EACH
Usually ships in 8 days or sooner.
at $100.00 USD/EACH
Call 1-888-358-4705 for availability.
at $520.00 USD/EACH
Usually ships in 11 days.
at $60.25 USD/EACH
In stock.
at $67.00 USD/EACH
In stock.
Replacement parts
Qty/Price
Availability
at $542.00 USD/EACH
Usually ships in 9 days.
at $542.00 USD/EACH
Usually ships in 26 days.
at $542.00 USD/EACH
Usually ships in 15 days.
at $72.00 USD/EACH
In stock.
at $72.00 USD/EACH
Usually ships in 15 days.
at $272.50 USD/EACH
In stock.
at $272.50 USD/EACH
Usually ships in 21 days.
at $272.50 USD/EACH
In stock.
at $542.00 USD/EACH
Usually ships in 17 days.
at $542.00 USD/EACH
Usually ships in 20 days.
at $665.00 USD/EACH
Usually ships in 7 days.
Extender; 3/4" tip
EW-04710-81
at $325.00 USD/EACH
Usually ships in 21 days.
Extender; 1" tip
EW-04710-82
at $335.00 USD/EACH
Usually ships in 18 days.
at $471.00 USD/EACH
Usually ships in 12 days.
at $272.50 USD/EACH
In stock.
Probe, 3-mm (1/8
EW-04712-12
at $272.50 USD/EACH
In stock.
4-element probe
EW-04712-30
at $1,396.00 USD/EACH
Usually ships in 56 days.
24-element probe
EW-04712-34
at $4,030.00 USD/EACH
Usually ships in 21 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title