log in or register
Cole-Parmer Chloride Analyzer, 120v
EW-02656-20
$6,892.00 USD/EACH
Usually ships in 6 days.
Qty
  • More accurate and precise than any other rapid method of salt determination!
  • Measurement repeatability ensures quality assurance and control
  • Compact design saves valuable bench space
  • Chloride standards are calibrated in both mg/L and mmol/L
This automatic chloride titrator provides accurate and highly reproducible measurement results. The system is ideal for use in the food, pharmaceutical, agricultural, and petroleum industries. The system utilizes a method based on coulometric titration—an absolute method where the reagent, silver ions, is precisely and quantitatively generated at the time of analysis by passing a constant current between electrodes. The end point is detected by the use of sensing electrodes which measure the change in the solution conductivity that occurs when excess silver ions are present in the solution. Unlike a conductivity meter, this analyzer actually titrates chloride with silver as in official methods.
Automatic features provide speed and simplicity, reducing labor costs and
Product Type Chloride Analyzer
Description Chloride analyzer
Reproducibility CV < 1% for 20 replicate samples at 200 mg/L
Accessories
Qty/Price
Availability
at $26.00 USD/EACH
In stock.
at $109.00 USD/PACKAGE
In stock.
at $109.00 USD/PACKAGE
In stock.
at $33.00 USD/EACH
Usually ships in 6 days.
at $16.75 USD/EACH
In stock.
at $40.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title