United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
Representative photo only
Cole Parmer Console drive 230 VAC 50 60 Hz
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-75211-15
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17050-01
NIST-Traceable Calibration for Peristaltic Pump Drive
$115.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Gear Pumps
>
Gear Pump Drives - For Micropump and Other Pump Heads
>
Cole-Parmer Variable-Flow Console Drives for MicroPump A-Mount Pump Heads
Share
|
Cole-Parmer Console drive; 230 VAC, 50/60 Hz
EW-75211-15
$890.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Permanent-magnet, direct-current (PMDC) motor is built for long-term applications
100:1 speed-control range controls flow over a wide range
Separate on-off switch maintains speed (flow) setting when drive is powered down
Maximum continuous-duty flow rates possible with Micropump
®
A-mount pump heads
Gradual, one-second acceleration for smooth flow rate changes
Simple variable speed control provides flow control for many fluid handling applications from filtration to chemical injection.
What's Included:
a 6-ft (1.8-m) modular line power cord; the 115 VAC model has a U.S. standard plug and the 230 VAC model has an IEC 320/CEE 22 connector (specify country to obtain correct plug).
Specifications
Temp range
32 to 104°F (0 to 40°C)
Duty cycle
Continuous
Motor type
PMDC
Power (Amps)
1.0
Power (Hz)
50/60
Dimensions
8-1/2"L x 6-3/8"W x 5-3/16"H
Motor (hp)
0.07
Power (VAC)
230
Brand
Cole-Parmer
Manufacturer number
75211-15
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration for Peristaltic Pump Drive
EW-17050-01
at $115.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Instruction Manual (398 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title