Cole-Parmer Essentials Digital Counter Pen
Representative image only.
Cole-Parmer Essentials Item # EW-20610-30
Combination marker and digital counter reduces errors—even when interrupted!
  • Automatically beeps every time a touch is made
  • Audio and visual counts to 99,999
  • Easy-to-read LCD
  • Felt-tipped pen with black water based ink is not permanent
  • Pen tip diameter of 1.397 mm
  • 1 year warranty
$25.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Width (InchShort)43/64
  • Battery1 392 Silver Oxide (included)
  • Depth (in)6
  • Colony Counter TypePen
  • DisplayLCD
  • MaterialABS Plastic
  • DescriptionDigital Counter Pen
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Includes
replaceable pen, cap, and one 392 silver oxide battery.
Key Features
  • Automatically beeps every time a touch is made
  • Audio and visual counts to 99,999
  • Easy-to-read LCD
  • Felt-tipped pen with black water based ink is not permanent
  • Pen tip diameter of 1.397 mm
  • 1 year warranty
More About this Item
Counter-Pen is a combination marker and digital counter designed for the lab and plant. Use it to count cell growth on petri dishes, inventory parts, and tally samples received. Counter beeps each time a mark is made ensuring an accurate tally. LCD simultaneously displays total and can be reset at the touch of a button. Durable ABS plastic pen features on/off switch and comes with cap.