United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
2 >
Representative photo only
Cole Parmer Digital Gauge 760 Torr 1 4 NPT M
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68936-80
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17040-12
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 0.75% or < 0.75%)
$140.00 USD/EACH/EACH
Usually ships in 4 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Gauges
>
Digital Pressure Gauges and Meters
>
Cole-Parmer 760 Torr Absolute Digital Vacuum Gauges
Share
|
Cole-Parmer Digital Gauge, 760 Torr; 1/4"; NPT(M)
EW-68936-80
$344.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Replace mercury manometers for monitoring vacuum systems
±0.25% test gauge accuracy
316 stainless steel wetted parts
Fast response—3 readings per second
Replace toxic mercury manomaters with these vacuum gauges! Use for monitoring laboratory or industrial vacuum manifolds, pumps, and systems. Large display provides better readability than mercury or liquid manometers. The 760 torr absolute scale measures atmospheric pressure when gauge port is open and reads zero at full vacuum.
Rugged epoxy-coated extruded aluminum case and rear cover, ABS/polycarbonate front bezel, front on/off push button, front accessible calibration adjustments.
Specifications
Product Type
Digital gauges
Media compatibility
316 stainless steel
Wetted materials
316 SS
Accuracy
±0.25% of full scale ±1 least significant digit
Unit of Measurement
torr
Resolution
1 torr
Range
760 to 0 torr absolute
Process connection
1/4" NPT(M)
Power
115 VAC/12 VDC power supply (included)
Case
aluminum
Display
3-digit LCD, 0.5"H
Brand
Cole-Parmer
Manufacturer number
CPARM760AD
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pressure / Vacuum (For gauges with accuracy 0.75% or < 0.75%)
EW-17040-12
at $140.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title