dummy
EW-28575-00
- Customize your own gel system depending on your needs and budget!
Mini Dual Gel Units. Small format is ideal for mini SDS-PAGE and mini agarose gel separations, plus it reduces the use of expensive reagents. These dual units let you run two agarose or polyacrylamide gels simultaneously under identical conditions; also compatible with most precast gels. The built-in cooling system ensures fast run times and flat, even banding patterns. Chamber gasket and clamps create a positive, leakproof seal.
Required System Components:
1) Gel Unit (order power supply separately)
For the Gel Wrap Gasket gel casting method, you will need:
2) Glass plate set(s)
3) Casting clamps (three per plate set for mini units)
Order the same thickness of:
4) GelWrap gasket (one per plate set)
5) Spacer set (one per plate set)
6) Combs (one per plate set)
What's Included:
cooling chamber with recirculation ports, CE-marked safety cover, power leads, and reservoir clamps. Combs, spacers and other gel casting accessories sold separately.
| Product Type |
Electrophoresis Vertical Systems |
| Buffer solution required (mL) |
250 |
| No. of wells |
10, 14 |
| Cell dimensions (" W) |
6.67 |
| Cell dimensions (" H) |
5 |
| Cell dimensions (" D) |
7.75 |
| Brand |
Cole-Parmer |
| Manufacturer number |
MGV-202-U |
at $4.25 USD/EACH
Call 1-888-358-4705 for availability.
at $30.00 USD/EACH
In stock.
at $30.00 USD/EACH
Call 1-888-358-4705 for availability.
at $30.00 USD/EACH
Call 1-888-358-4705 for availability.
at $30.00 USD/EACH
Call 1-888-358-4705 for availability.
at $30.00 USD/EACH
In stock.
at $30.00 USD/EACH
In stock.
at $30.00 USD/EACH
In stock.
at $30.00 USD/EACH
In stock.
at $18.50 USD/EACH
Call 1-888-358-4705 for availability.
at $18.50 USD/EACH
In stock.
at $18.50 USD/EACH
In stock.
at $18.50 USD/EACH
In stock.
at $7.50 USD/EACH
In stock.
at $7.50 USD/EACH
Call 1-888-358-4705 for availability.
at $7.50 USD/EACH
In stock.
at $7.50 USD/EACH
In stock.
at $23.00 USD/EACH
In stock.
at $1,116.00 USD/EACH
Usually ships in 7 days.
at $2,042.00 USD/EACH
Usually ships in 30 days.
at $1,845.00 USD/EACH
Usually ships in 30 days.
at $547.00 USD/EACH
In stock.
at $619.00 USD/EACH
Usually ships in 8 days.
at $1,500.00 USD/EACH
Usually ships in 20 days.
at $1,532.00 USD/EACH
Usually ships in 30 days.
at $2,695.00 USD/EACH
Usually ships in 30 days.
at $2,574.00 USD/EACH
Usually ships in 30 days.
at $2,709.00 USD/EACH
Usually ships in 35 days.
at $2,656.00 USD/EACH
Usually ships in 55 days.
Technical Resources
Related technical articles (Roll over for summary)
Show All...
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334