Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Cole-Parmer Electrode Support Clamp, nickel-plated zinc
Item# EW-08027-06
$61.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 12 days.
Qty
  • Provide support for your specific laboratory equipment
This clamp is designed to supports your electrode with an electrode holder at the desired level and angle. Integrated connector included.
Product Type Specialty Clamps
Description Electrode support clamp with built in holder
Dimensions (" L) 7 in (17.8 cm)
Material (Clamp) Nickel-plated Zinc
Model 916079
Qty/ea 1
Brand Cole-Parmer
Technical Resources
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456