Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Cole-Parmer Flowmeter System, 65-mm Direct-Reading Flowtube for Air; 1 LPM
Item# EW-32047-09
$46.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 11 days.
Qty
Product Type Variable Area Flowmeter
Description Variable Area, Direct Reading Flowmeter
Max flow rate (LPM) 1
Material (Flow Tube) Borosilicate
Material (float) Glass
Max operating temperature (º F) 250
Max operating temperature (º C) 121
Max pressure 200 psi
Accuracy ±2% of full scale
Process connection 1/8" NPT(F)
Qty/ea 1
Repeatability ±0.25% full-scale
Brand Cole-Parmer
Calibrations
Qty/Price
Availability
at $246.00 USD/EACH
Usually ships in 14 days.
Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456