United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
Representative photo only
Cole Parmer Hot Plate Ceramic top 12 x 12 115 VAC 60 Hz
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-04801-14
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17002-26
Innocal NIST Calibration for Hotplate Only
$130.00 USD/EACH/EACH
Usually ships in 15 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Heating Equipment
>
Hot Plates
>
Analog Hot plates
>
Analog Ceramic Top Hot Plates
>
Cole-Parmer Ceramic Top Hot Plates
Share
|
Cole-Parmer Hot Plate, Ceramic top, 12" x 12", 115 VAC, 60 Hz
EW-04801-14
$510.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Maximum temperature reached in approximately 2 minutes
Epoxy-coated cast-aluminum body is designed for durability and stability. A corrosion-resistant ceramic top is perfect for your high-temperature applications; ceramic top is housed in an aluminum frame for cooler edges.
What's Included:
6-ft (1.8-m) cord with plug.
Specifications
Top plate dimensions (in.)
12 x 12
Top plate material
Ceramic
Temp range (º F)
120 to 842°F
Temp range (°C)
49 to 450°C
Dimensions
12 W x 4 7/8 H x 15 D
Max load capacity
25 lb
Power (VAC)
115
Power (Hz)
60
Power (Amps)
13
Power (watts)
1450
Brand
Cole-Parmer
Calibrations
Qty
/
Price
Availability
Innocal NIST Calibration for Hotplate Only
EW-17002-26
at $130.00 USD/EACH
Usually ships in 15 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Keeping Samples the Right Temperature
Stirrers and Hot Plates Selection Guide
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title