United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 20
of
24 >
Representative photo only
Cole Parmer Kynar Gas Sampling Bag 12 x 12 On Off and Septum valves
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-01410-52
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Forensics
>
Safety & Response Products
>
Air Sampling and Gas Analysis
>
Air Sampling
>
Sampling Bags
>
Cole-Parmer Kynar Gas Sampling Bags
Share
|
Cole-Parmer Kynar Gas Sampling Bag, 12" x 12", On/Off and Septum valves
EW-01410-52
$52.00 USD/EACH
Quantity Discounts
Order
Discount
Price
[X]
10
10%
$46.80/EACH
50
15%
$44.20/EACH
100
20%
$41.60/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Corrosive-resistant Kynar for all your applciation needs
Strong seams for added durability
All standard models have either an on/off valve (3⁄16'' nickel-plated brass hose barb) or a silicone septum in polypropylene housing; dual-valve models have both. Max temp: 160°F (71°C). Individually sealed in a dust-protected outer bag that also blocks out sunlight.
Standard Kynar Bags
Single-wide seam; available in 2-mil and 4-mil thick styles.
Dual-Valve Kynar Bags
Double-wide seamed bags have both on/off and septum valves for convenient mixing.
Dual-Valve Kynar Bags
Double-wide seamed bags have both on/off and septum valves for convenient mixing.
Specifications
Product Type
Sampling Bag
Thickness
3-mil
Valve
Dual valve, on/off and septum
Material
PVDF
Volume (Liters)
3.8
Dimensions (" W)
12
Dimensions (" L)
12
Brand
Kynar
Manufacturer number
01410-52
Model
01410-52
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
Isolating Genomic DNA from Whole Blood
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title