United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 5
of
8 >
Representative photo only
Cole Parmer LabGEN 125 Homogenizer Kit Hard tissue version
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Brochure
Related technical articles
Service & Support
Related media
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-04727-11
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Mixers and Blenders
>
Homogenizers
>
Cole-Parmer Homogenizers
>
Cole-Parmer Labgen Tissue Homogenizers
>
Cole-Parmer LabGEN Tissue Homogenizers
Share
|
Cole-Parmer LabGEN 125 Homogenizer Kit, Hard tissue version
EW-04727-11
$980.00 USD/EACH
Usually ships in 3 days.
Qty
:
Add to Cart
Save to shopping list
Handle small tissue samples, PCR, RNA, and DNA up to heavy-duty tissue processing and cell disruption
Variable-speed motor performs up to 30,000 rpm
Homogenize tissue samples quickly and efficientlywithout worrying about cross-contamination
Cole-Parmer’s LabGEN homogenizers are well-suited for a wide range of processing applications —use as a handheld unit or post-mount to the optional stand for hands-free operation. Use these homogenizers with either the stainless steel rotor-stator generators or the plastic tip probes (tip adapter 04727-88 required). Order components separately.
Plastic tip probes are maintenance-free and do not require the disassembly associated with traditional stainless steel generators. Tips are available in both soft tissue (deaggregations) and hard tissue (frozen) versions; designed to work with sample volumes between 0.25 to 30 mL. Engineered from durable plastics, these tips may be autoclaved and reused, or disposed of after each use to eliminate cross contamination.
Labgen 125 Homogenizing Kits
are ideal for RNA, DNA, and PCR applications. They contain everything you need to begin homogenizing tissue samples—a Labgen 125 homogenizer, 12 plastic tip probes, tip adapter 04727-88, and a storage case.
Specifications
Product Type
Homogenizer
Max sample size (mL)
30
Speed (rpm)
5000 to 30,000
Generator included
Yes - 12 plastic tip probes for hard tissue
Brand
Cole-Parmer
Power (VAC)
115
Power (Hz)
50/60
Watts
125
Accessories
Qty
/
Price
Availability
Cole-Parmer LabGEN Homogenizer stand for 04727-series
EW-04727-89
at
$340.00 USD/EACH
In stock.
Add to Cart
Additional Motor Support Rod for Cole-Parmer LabGEN Homogenizer 125 and 700
EW-04727-90
at
$65.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Cole-Parmer
LabGEN
Homogenizer Brochure (438 KB)
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
The Use of Steel-Tip Homogenizer Probe Results in Sample Carryover Contamination
Smart Mixing
Mixer Selection Guide
Show All...
Positioning a Mixer in a Cylindrical Vessel
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Related media
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title