United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 7
of
9 >
Representative photo only
Cole Parmer Polystat Standard Plus digital immersion circulator with bridge 115V 60 Hz
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Instruction manual
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-12120-11
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17001-13
NIST-Traceable Calibration; Temperature Baths
$250.00 USD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Baths and Circulators
>
Immersion Circulators
>
Cole-Parmer Polystat Immersion Circulators
Share
|
Cole-Parmer Polystat Standard Plus digital immersion circulator with bridge; 115V, 60 Hz
EW-12120-11
$1,123.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
High-stability immersion circulators feature low-level sensor for temperature control up to 200°C
PID control with easy-to-use LCD display in four languages and up to five temperature setpoints
Pump connections for circulation to external equipment in open- or closed-loop applications
Resolution of 0.01° reads out in °C, °F, or °K
The immersion circulator is designed with an easy-to-read LCD in your choice of temperature units from Celsius/Farrenheit/or Kelvin. Programmable high/low/limit set points on easy-to-use PID temperature controller. Adjustable pump nozzle.
Standard
and
Standard Plus Controllers
feature five programmable set points, a two-speed pump, and an adjustable pump nozzle.
What's Included:
6-ft (1.8-m) power cord with plug, M16 x 1 connection and adapters for included 8- and 12-mm hose barbs with clamps.
Specifications
Product Type
Circulators (Immersion)
Min temperature (º C)
Ambient +10
Max temperature (º C)
150
Dimensions (" W)
4.4
Dimensions (" H)
12.6
Dimensions (" D)
8.1
Temp control
PID
Temp sensor
Pt RTD
Immersion depth
5-3/4" (14.6 cm)
Low-level cutoff
Yes
Pressure pump (Max head at 0 flow)
4.5 psi (310 mbar)
Wetted materials
stainless steel; polymers include: Polyvinylidenefluoride/hexafluoropropene (Viton®), ethylene propylene diene monomer (EPDM); polyphenylene sulfide (PPS); Polyetherimide (PEI); liquid crystal polymer (Vectra®)
Heater wattage
1 kW
Pump type
force
Stability
+/- 0.05°C
Suction pump
No
Max flow rate (Liters/min)
17
Power (VAC)
115
Power (Hz)
60
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Temperature Baths
EW-17001-13
at $250.00 USD/EACH
Usually ships in 21 days.
Add to Cart
Technical Resources
Instruction Manual
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Bath and Circulator Specifications Terminology
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title