United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
16 >
EW-13042-01
Cole-Parmer
®
Polystat
®
recirculator, 17 lpm, 250W cooling capacity, 115V 60 Hz
Click to enlarge
More Product Details
Specifications
Calibrations
Reconditioned & Like New
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-13042-01
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17001-13
NIST-Traceable Calibration; Temperature Baths
$250.00 USD/EACH/EACH
Usually ships in 21 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Baths and Circulators
>
Recirculators & Chillers
>
Cole-Parmer Polystat Cooling/Heating Laboratory Recirculators
Share
|
Cole-Parmer Polystat recirculator, 17 lpm, 250W cooling capacity, 115V 60 Hz
EW-13042-01
$2,385.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Full-range cooling for quick ramp up and cool-downsaves valuable time
Robust cooling and heating from -10 to 80°C
Compact design maximizes bench space and provides quick temperature changes
Internal tank design minimizes fluid evaporation
True thermostatic control laboratory chiller removes or adds temperature as required by your application. This closed circulator is ideal for low load applications such as cooling analytical instrumentation, jacketed reactors, and rotary evaporators. Use as an economical chiller to replace tap water cooling in small laboratory applications. Powerful force/suction pump provides flow rates up to 21 L/min at pressures up to 11.7 psi (805 mbar). Features overfill port and liquid-level switch that can either trip audible alarm or fault machine. Include USB port.
What's Included:
6-ft (1.8-m) power cord with 3-prong plug, funnel, and 8- and 12-mm hose barb fittings with clamps for external connections.
Specifications
Product Type
Circulators (Cooling/Heating)
Capacity (Liters)
2.8
Min temperature (º C)
-10
Max temperature (º C)
80
Dimensions (" W)
9
Dimensions (" H)
25
Dimensions (" L)
16.5
Temp stability
+/- 0.1°
Cooling capacity (at 20° C)
250 W
Temp setting
digital
Low-level cutoff
Yes
Heater wattage
1200
Pump type
force
Max pump pressure (psi)
4.4 psi
Max pump pressure (mbar)
300 mbar
Power (VAC)
115
Power (Hz)
60
Flow rate
17 L/min
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Temperature Baths
EW-17001-13
at $250.00 USD/EACH
Usually ships in 21 days.
Add to Cart
Reconditioned & Like New
Qty
/
Price
Availability
LIKE NEW 13042-01
EW
-
00282FW
at
$1,929.50 USD/EACH
In stock.
Add to Cart
RECOND 13042-01
EW
-
00304SR
at
$1,454.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Keeping Samples the Right Temperature
Chiller/Recirculator Selection Guide
Learn how to choose the chiller that best suits all of your cooling needs
Show All...
Bath and Circulator Specifications Terminology
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title