Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Cole-Parmer Polyurethane Tubing, aqua translucent, 1/16" x 1/8", 100'
Item# YO-95868-00
$54.00 USD/PACKAGE
Quantity Discounts
Qty:
Price:
4
$48.60
10
$45.90
20+
$43.20
    Make your lines easily identifiable by color-coding them! This ether-grade polyurethane is an an economical solution that offers superior heat resistance, low compression set, good elongation and recovery. It is odorless and nontoxic and has excellent biocompatibility. Ether-based polyurethane is a good solution for weak organic acids/oils. Sterilize with ethylene oxide.
    Use green, blue, red and black tubing from –70 to 185° F (–57 to 85° C); our aqua-tinted polyurethane is usable from –40 to 180° F (–42 to 82° C).
    Working pressure (psi) 100
    Maximum temperature (º F) 185
    Minimum temperature (º F) -70
    Tubing ID (in.) 0.0625
    Tubing ID (mm) 1.6
    Tubing OD (in.) 0.03125
    Tubing OD (mm) 0.8
    Max psi 100
    Translucent Yes
    Sterilizable Yes
    Color Translucent
    Feet per pack 100
    Tubing formulation Polyurethane
    Product Type Tubing
    Brand Cole-Parmer
    Manufacturer number AP01T061PEBLT
    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Do you have questions about this product?
    get answers from real customers and in-house experts with AnswerBox.
    1 Question | 1 Answer
    Displaying question 1
    • Q:
      Does this tubing meet USP Class VI standard?
      Asked on 6/29/2011 by Mike from Minneapolis, MN
      1 answer
      • Staff Reviewer
        A:
        No, this has no certifications (USP, FDA, NSF, etc).
        Answered on 6/29/2011 by Jen from Technical Support
    Displaying question 1
    Do you have a question about this product? 
    Product Reviews
    Customers Who Bought This Item Also Bought