dummy
EW-59824-00
Call 1-888-358-4705 for availability.
Qty:
- Analyze protein, oil, and moisture content anywhere!
- Displays results in less than one minute
Enhance the profitability of your grains and dairy products by ensuring their optimum protein, oil, and moisture contents. Analyzer is designed for rugged use—take with you on the plant floor or in the field.
Meter analyzes protein and oil both on an “as-is” basis and a constant moisture (CM) basis. Simply pour sample into sample holder and place holder into the meter. Results are displayed in less than one minute. Download data to your 486 computer or better with the included calibration software and RS-232 cable.
Includes calibration for HRS wheat. When ordering, specify one additional calibration from the following: HRW, protein and moisture in SWW, durum wheat, SRW, 2 row barley, 6 row barley, oats, corn, soybean, canola, mustard, flax, milo, or long- and medium-grain rough rice. Oil calibration is included with corn, soybean, canola, mustard, and flax calibrations.
What's Included:
analyzer, 16-mm sample cup for general analysis, liquid/slurry sample holder, light shield, calibration software, RS-232 cable, and six AA batteries. Order additional factory-set calibrations and other accessories separately.
| Product Type |
Digestion Spectroscopes |
| Optical range (AU) |
0 to 6 |
| Resolution (AU) |
0.0001 |
| Range (%) |
1.0 to 95 |
| Spectrum range |
12 filters covering wavelengths from 893 to 1045 nm |
| Output |
RS-232 via included 25-pin connector with 6-ft cable |
| Resolution |
0.0001 AU |
| Sample size |
approximately 100 grams |
| Stability (Mili-AU) |
0.05 |
| Stability |
0.05 Mili-AU |
| Scan time |
usually less than one minute |
| Display |
LCD |
| Power (VAC) |
120 or 220 adapter |
| Battery |
1.5 V AA |
| Brand |
Cole-Parmer |
at $7.50 USD/PACKAGE of 4
In stock.
at $300.00 USD/EACH
Call 1-888-358-4705 for availability.
at $595.00 USD/EACH
Call 1-888-358-4705 for availability.
at $295.00 USD/EACH
Call 1-888-358-4705 for availability.
at $55.00 USD/EACH
Call 1-888-358-4705 for availability.
at $55.00 USD/EACH
Call 1-888-358-4705 for availability.
at $120.00 USD/EACH
Call 1-888-358-4705 for availability.
at $420.00 USD/EACH
Usually ships in 14 days.
at $50.00 USD/BOX of 10
Usually ships in 14 days.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334