dummy
EW-44175-04
Call 1-888-358-4705 for availability.
Qty:
- Chill, freeze, heat, or mix biological samples at your work station
- Store 5 programs in memory for precisely timed and repeatable routines
- Choose blocks for centrifuge tubes, assay plates, PCR tubes & plates, and more
- Two-block unit offers two different or same temperatures
Replace your ice buckets or inconvenient freezer blocks with this compact, economical benchtop chilling/heating block. Features include a two-line LCD that reads actual and set point temperatures, 30-day countdown timer with alarm and auto/off, built-in data logger, and RS-232 I/O port for bidirectional communication with an IBM® compatible computer. Energy-efficient Peltier design chills or heats biological samples from -10 to 100°C with ±1°C digital control.
Programmable Orbital Mixing Chilling/Heating Block is fully programmable with built-in orbital mixer. This model can store 5 programs in memory of as many as 10 steps per program where each step can be a temperature, time and mixing speed. Each program can be repeated from 1 to 99 times automatically.
What's Included:
Chiller/heater module, power cord, instruction manual, and a 12 VDC power supply. Order cover and aluminum blocks separately.
| Product Type |
Chilling/Heating Blocks |
| Description |
Programmable Orbital Mixing Chilling/Heating Block |
| Dimensions (overall) |
8-3/4" L x 6-1/2" W x 3-1/2" H (22.2 x 16.5 x 8.9 cm) |
| Power (VAC) |
100 to 230 |
| Stability |
±1°C |
| Temp display |
Two line LCD |
| Temp range |
-10 to 100°C |
| CE Compliance |
Yes |
| Brand |
Cole-Parmer |
| Manufacturer number |
SC25CP |
at $161.00 USD/EACH
Usually ships in 15 days.
at $176.00 USD/EACH
In stock.
at $231.00 USD/EACH
Usually ships in 12 days.
at $221.00 USD/EACH
Call 1-888-358-4705 for availability.
at $186.00 USD/EACH
Usually ships in 9 days.
at $151.00 USD/EACH
Usually ships in 8 days.
at $156.00 USD/EACH
Usually ships in 7 days.
at $55.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334