Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Cole-Parmer Sabouraud Dextrose Agar, 500 g

EW-14202-62
$56.00 USD/EACH

Usually ships in 6 days.
Qty

  • Media cultures are tested and standardized to ensure consistent and reliable performance
  • Available for sale in the United States only
  • For lab use only; not intended for in vitro purposes
Use this USP medium to perform total combined yeast and mold counts (microbial limit tests).

Offered in the most popular formulations, our media is thoroughly tested and subject to strict examination and standardization to ensure reliable and consistent lot-to-lot performance. Contact one of our Application Specialists to find out the availability of other dehydrated culture media and learn about our custom blending service.

Each order includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and a shelf-life guarantee. Wide mouth openings on bottles let you scoop out powder efficiently without dusting the countertop. The lightweight plastic bottles have indented hand grips for steady handling. Wide, flat cap allows you to stack containers and save space. Media are formulated to dissolve quickly.
Product Type Agar
Capacity (g) 500
Media Name Media S-U
Qty/pk 1
Brand Cole-Parmer
Manufacturer number 14202-62





Technical Resources
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title