Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
Sorry, this item is now discontinued.browse similar itemsor Contact us for more information.

Cole-Parmer Safetypette Pipette Filler, fits 1-100 mL pipettes

Item# EW-25200-30

$160.00 USD/EACH

Discontinued
Qty

  • Egg-shaped bulb provides a comfortable grip and fast suction for both right- and left-handed users
  • Powerful suction quickly draws fluids
  • Filter protects bulb from damage by caustic liquids and filters air to prevent cross contamination of pipettes.
Uniquely designed, lightweight pipette filler fits comfortably in either hand enabling smooth, variable speed control when aspirating or dispensing. Simple thumb-button operation provides precise control. Safetypette works with any pipette between 1 mL and 100 mL, and is compatible with both blow-out and to-deliver pipettes. Autoclavable.
What's included: integral 0.45 µm filter, silicone pipette holder, and three silicone colored pipette cones for safety color coding.
Product Type Pipetting Accessory
Color three silicone colored pipette cones
Qty/ea 1
Brand Cole-Parmer
Manufacturer number 475-165





Technical Resources
Virtual Catalog (PDFs)
  • Catalog - Spanish
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews

    REVIEW SNAPSHOT®

    by PowerReviews
    Cole-ParmerSafetypette pipette filler
     
    3.0

    (based on 2 reviews)

    Reviewed by 2 customers

    Displaying reviews 1-2

    Back to top

     
    5.0

    Pipette filler

    By JC

    from Malibu, Ca

    About Me Academic/Professional Researcher

    Verified Reviewer

    Pros

    • Accurate
    • Clear Markings
    • Easy To Use
    • Sturdy

    Cons

      Best Uses

      • In The Lab

      Comments about Cole-Parmer Safetypette pipette filler:

      I use the pipette for various titrations that do not require a micropipette

       
      1.0

      sucks or does it

      By haha

      from Denver, CO

      About Me Lab Assistant

      Pros

      • Easy To Use

      Cons

      • Breaks Easily
      • Inaccurate

      Best Uses

      • In The Lab

      Comments about Cole-Parmer Safetypette pipette filler:

      breaks really easily.

      Displaying reviews 1-2

      Back to top

      Customers Who Bought This Item Also Bought